Learn More
TMC Hallcrest™ Thermax™ 5 Level Clock Temperature Indicators
Provide a fast, simple temperature monitoring system based on proprietary color change technology. TMC Hallcrest™ Thermax™ 5 Level Clock Temperature Indicators record the highest temperature achieved with irreversible color change.
Brand:  TMC Hallcrest™ M05CTHERG4D01P1
Product Code. 11760879
  • 17.15€ / Pack of 10
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Heat-sensitive round label to monitor sterilization temperature
  • Inexpensive and easy to use
  • Adhesive
  • Immediate response.
  • Permanent color change records highest temperature achieved.
  • Oil, water and steam resistant.
  • Accuracy tolerances: 29-99°C (+/-1°C) / 100 -154°C (+/-1.5°C) 160-290°C (+/-1°C)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
TMC Hallcrest™ Thermax™ 5 Level Clock Temperature Indicators > Temperature Range: 116 to 138°C