Learn More
MP Biomedicals™ TallPrep™ Metal Bead Lysing Matrix Tubes
TallPrep™ Metal Bead Lysing Matrix Tubes for nucleic acid isolation and purification
Brand: MP Biomedicals™ 116945050
This item is not returnable.
View return policy
Description
- Each impact-resistant 4.5 mL tube contains twelve 3.175 mm (1/8 in.) diameter stainless steel beads
- Use to break hard/brittle samples
- Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing
Specifications
Stainless Steel | |
Store at Room Temperature. | |
DNase Free | |
Nucleic Acid isolation and purification | |
Lysis Matrix Tube | |
RNase Free |
4.5 mL | |
Animal Tissue, Bacteria (Gram +/-), Bone, Feces, Fixed Samples, Food, Fungi, Heart, Lung, Plants, Roots, Seeds, Skeletal Muscle, Sludge, Soil, Spores, Wastewater, Yeast | |
FastPrep-24, FastPrep 120, FastPrep-96 | |
Stainless Steel | |
50 x 4.5 mL |