missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ TallPrep™ Metal Bead Lysing Matrix Tubes

TallPrep™ Metal Bead Lysing Matrix Tubes for nucleic acid isolation and purification

Brand:  MP Biomedicals™ 116945100

 View more versions of this product

Product Code. 15469744

  • 525.00€ / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Each impact-resistant 4.5 mL tube contains twelve 3.175 mm (1/8 in.) diameter stainless steel beads
  • Use to break hard/brittle samples
  • Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stainless Steel
Store at Room Temperature.
DNase Free
RNase Free
4.5 mL
Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores, Food, Plants, Roots, Fixed Samples, Tough Tissues, All Samples
100 x 4.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.