Learn More
Lenz Laborglasintrumente Table Top Reaction Units, Lab Scale
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
2730.00€ - 3305.00€
Specifications
Material Duran Glass
View More Specs
Products 6
Product Code Brand Capacity (Metric) Includes Price Quantity & Availability  
Product Code Brand Capacity (Metric) Includes Price Quantity & Availability  
11903831
View Documents
Lenz Laborglasintrumente
06260270
1 L Lab Flange 100 LF
3280.00€
Each
11913591
View Documents
Lenz Laborglasintrumente
06260273
2 L Lab Flange 100 LF
3305.00€
Each
11963821
View Documents
Lenz Laborglasintrumente
06260137
100 mL Lab Flange 60 LF
2730.00€
Each
11973821
View Documents
Lenz Laborglasintrumente
06260149
250 mL Lab Flange 60 LF
2740.00€
Each
11983821
View Documents
Lenz Laborglasintrumente
06260158
500 mL Lab Flange 60 LF
2755.00€
Each
11993821
View Documents
Lenz Laborglasintrumente
06260258
500 mL Lab Flange 100 LF
3250.00€
Each
Description
Description
  • For stirring under normal pressure or vacuum, with possibility of addition of liquids and boiling under reflux.
  • High-quality system components: Reaction vessels (100, 250, 500, 1000 or 2000 ml) with thermostatic jacket (heatable by a circulation bath), with bottom discharge valve (PTFE, 10 mm, no dead volume), made of DURAN™ tubing
  • Flat flange lid (LF 60 or LF 100) with center neck and two angled side necks, material: DURAN™
  • Funnels, cylindrical (100 or 250 ml) with graduation, made of DURAN™ tubing Reflux condensers (details see on page 71), made of DURAN™ tubing
  • Stirrer guide and stirrer with 10 mm stirrer shaft and lateral PTFE blade
Specifications
Specifications
Duran Glass