Learn More
Lenz Laborglasintrumente Table Top Reaction Units, Lab Scale
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
2730.00€ - 3305.00€
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | ||||
11903831
|
Lenz Laborglasintrumente
06260270 |
1 L | Lab Flange 100 LF |
3280.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11913591
|
Lenz Laborglasintrumente
06260273 |
2 L | Lab Flange 100 LF |
3305.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11963821
|
Lenz Laborglasintrumente
06260137 |
100 mL | Lab Flange 60 LF |
2730.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11973821
|
Lenz Laborglasintrumente
06260149 |
250 mL | Lab Flange 60 LF |
2740.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11983821
|
Lenz Laborglasintrumente
06260158 |
500 mL | Lab Flange 60 LF |
2755.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11993821
|
Lenz Laborglasintrumente
06260258 |
500 mL | Lab Flange 100 LF |
3250.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For stirring under normal pressure or vacuum, with possibility of addition of liquids and boiling under reflux.
- High-quality system components: Reaction vessels (100, 250, 500, 1000 or 2000 ml) with thermostatic jacket (heatable by a circulation bath), with bottom discharge valve (PTFE, 10 mm, no dead volume), made of DURAN™ tubing
- Flat flange lid (LF 60 or LF 100) with center neck and two angled side necks, material: DURAN™
- Funnels, cylindrical (100 or 250 ml) with graduation, made of DURAN™ tubing Reflux condensers (details see on page 71), made of DURAN™ tubing
- Stirrer guide and stirrer with 10 mm stirrer shaft and lateral PTFE blade
Specifications
Duran Glass |