Learn More
Lenz Laborglasintrumente Table Top Reaction Units, Lab Scale
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 06260149
This item is not returnable.
View return policy
Description
- For stirring under normal pressure or vacuum, with possibility of addition of liquids and boiling under reflux.
- High-quality system components: Reaction vessels (100, 250, 500, 1000 or 2000 ml) with thermostatic jacket (heatable by a circulation bath), with bottom discharge valve (PTFE, 10 mm, no dead volume), made of DURAN™ tubing
- Flat flange lid (LF 60 or LF 100) with center neck and two angled side necks, material: DURAN™
- Funnels, cylindrical (100 or 250 ml) with graduation, made of DURAN™ tubing Reflux condensers (details see on page 71), made of DURAN™ tubing
- Stirrer guide and stirrer with 10 mm stirrer shaft and lateral PTFE blade
Specifications
Lab Flange 60 LF | |
Duran Glass | |
250 mL |