Learn More
Thermo Scientific™ T4 DNA Ligase (5 U/μL)
T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA.
47.79€ - 322.00€
Specifications
Concentration | 5 U/μL |
---|---|
Enzyme | DNA Ligase |
Compatible Buffer | 10X T4 DNA Ligase Buffer |
Product Type | T4 DNA Ligase |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10723941
|
Thermo Scientific™
EL0011 |
1,000 units |
82.75€
1000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10621441
|
Thermo Scientific™
EL0012 |
5 x 1,000 units |
322.00€
5000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10339509
|
Thermo Scientific™
EL0014 |
200 units |
47.79€
200 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Catalyze the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA with the Thermo Scientific™ T4 DNA Ligase. The enzyme repairs single-strand nicks in duplex DNA, RNA, or DNA/RNA hybrids. It also joins DNA fragments with either cohesive or blunt termini, but has no activity on single-stranded nucleic acids.
The T4 DNA Ligase requires ATP as a cofactor.
Highlights
- Active in Themo Scientific restriction enzyme, PCR, and RT buffers (when supplemented with ATP)
- Fast – sticky-end ligation is completed in 10 minutes at room temperature
- Supplied with PEG solution for efficient blunt-end ligation
Applications
- Cloning of restriction enzyme generated DNA fragments
- Cloning of PCR products
- Joining of double-stranded oligonucleotide linkers or adaptors to DNA
- Site-directed mutagenesis
- Amplified fragment length polymorphism (AFLP)
- Ligase-mediated RNA detection (see Reference 3)
- Nick repair in duplex DNA, RNA or DNA/RNA hybrids
- Self-circularization of linear DNA.
Notes
- Binding of T4 DNA Ligase to DNA may result in a band shift in agarose gels. To avoid this, incubate samples with 6X DNA Loading Dye & SDS Solution at 70°C for 5 min or 65°C for 10 minutes and chill on ice prior to electrophoresis.
- The volume of the ligation reaction mixture should not exceed 10% of the competent cell volume in the transformation process.
- Prior to electro-transformation, remove T4 DNA Ligase from the ligation mixture by spin column or chloroform extraction. The extracted DNA can be further precipitated with ethanol.
Specifications
5 U/μL | |
DNA Ligase | |
10X T4 DNA Ligase Buffer | |
T4 DNA Ligase |