Learn More
Prestige Medical T-Classic Media Autoclave
T-Classic Media Autoclave, 12 L benchtop autoclave, 121°C cycle, with european plug.
Brand: Prestige Medical 210048XP
Description
T-Classic Media Autoclave, 12 L benchtop autoclave, 121°C cylce, european plug
- Lightweight and portable
- 12 liter capacity
- Sterilizes at 121°C
The Classic is reliable and trusted worldwide, designed to give you the most cost effective sterilization solution. It is simplicity itself to use, fully automatic with a simple, one touch operation and guaranteed to meet your needs.
Specifications
2.6 gal. | |
270 mm | |
10.8 in. | |
13.6 in. | |
Autoclave | |
121°C |
12 L | |
8.4 in. | |
210 mm | |
340 mm | |
249.8°F | |
Non-sterile |