missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios Surfa'Safe Premium Foaming Spray

Disinfecting detergent foam for cleaning and disinfection of surfaces, external equipment structures and non-submersible and non-invasive medical devices.

Brand:  Laboratoires Anios 2419544


Product Code. 15441535

  • 19.90€ / 750mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Detergent disinfectant foam for surfaces and medical devices

  • Cleans and disinfects in a single operation
  • Without CMR, without alcool, without perfume nor colorant
  • Broad spectrum antimicrobial and optimised contact time
  • Compact foam dispenser to limit the formation of aerosols and
    volatile components (750ml)
  • Formula ensures compatibility with a wide range of materials,
    including polymers (e.g.: polycarbonate, plexiglas...)
  • Dense foam for better surface adhesion (750ml)
  • Surfaces dry fast without marks
  • Type 4 biocide which can be used on surfaces which come into
    contact with food products, followed by rinsing
  • Improved ecotoxicological profile: GHG*
    emissions reduced by 7% relative to a similar product
  • pH : 6 ± 0.5
Specifications

Specifications

Foaming Disinfectant
For Cleaning and Disinfection of Surfaces, External Structures of Equipment
12/Pk.
750 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : didecyldimethylammonium chloride, amines, N-C12-14-alkyltrimethylenediamines

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Laboratoires Anios Surfa'Safe Premium Foaming Spray > Volume: 750mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.