missing translation for 'onlineSavingsMsg'
Learn More
Sulfasalazine, 97%, Thermo Scientific Chemicals Product Code.: 15710909

Sulfasalazine, 97%, Thermo Scientific Chemicals

Product Code. 15710909
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Product Code. 15710909

Brand: Thermo Scientific Acros 461240250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-79-1
Molecular Formula C18H14N4O5S
Molecular Weight (g/mol) 398.39
InChI Key OQANPHBRHBJGNZ-BKUYFWCQSA-N
IUPAC Name (3Z)-6-oxo-3-(2-{4-[(pyridin-2-yl)sulfamoyl]phenyl}hydrazin-1-ylidene)cyclohexa-1,4-diene-1-carboxylic acid
SMILES OC(=O)C1=C\C(C=CC1=O)=N/NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1

Specifications

Melting Point 260.0°C to 265.0°C
Assay Percent Range 96.0 % min. (HPLC)
Quantity 25 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: slightly soluble in alcohols, insoluble in ether, benzene, chloroform
Formula Weight 398.39
Chemical Name or Material Sulfasalazine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0456968 Certificate TypeCertificate of Analysis Date13/10/2023 Catalog Number461240050,461240250,461241000
Lot NumberA0451247 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number461240050,461240250,461241000
Lot NumberA0374705 Certificate TypeCertificate of Analysis Date15/09/2021 Catalog Number461240050,461240250,461241000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfasalazine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.