Learn More
Cole-Parmer™ Stuart™ Receiving Flask
Brand:  Cole-Parmer™ 10096-69
Product Code. 12009131
  • 330.00€ / Each
Estimated Shipment: 07-07-2025
to see stock.
Explore more special offers
Specifications
1000 mL
All glassware sets, S35 spherical socket
Product Title
Cole-Parmer™ Stuart™ Receiving Flask > Capacity: 1000mL