Learn More
MP Biomedicals™ Striker Bolt For FastPrep-24™
Striker Bolt is a part for the FastPrep-24™ 5G.
Brand: MP Biomedicals™ 116005508
Description
- Samples and buffers are added to a 2.0 mL Lysing Matrix tube containing specialized Lysing Matrix beads.
- The ergonomic design of the instrument ensures an easy loading of the sample tubes that remains securely sealed during the processing.
- The homogenization speed and duration times are manually programmed or accessed from onboard memory.
- After setting your speed and time with the touch of a button, just push run, and in less than a minute your samples are completely lysed.
Specifications
FastPrep-24 5G | |
1 Ea. |
Product Content Correction
Product Title
MP Biomedicals™ Striker Bolt For FastPrep-24™ > Striker Bolt; For use with: FastPrep-24 5G
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.