Learn More
IBA Lifesciences Strep-tag™II Peptide
Strep-tag™II peptide is a synthetic peptide recognized by Strep-Tactin™ and Strep-Tactin™XT
Brand:  IBA Lifesciences 2-1018-002
Additional Details : Weight : 0.00900kg
Product Code. 12813216
  • 93.25€ / 1.80mg
Estimated Shipment: 12-07-2024
to see stock.
Add to basket
Description
Description
  • Synthetic Strep-tag™II peptide recognized by Strep-Tactin™, Strep-Tactin™XT as well as the monoclonal antibodies, StrepMAB-Classic and StrepMAB-Immo.
  • It can be used for competitive elution of Strep-tag™II fusion proteins or as control protein in dot blot assays.
Specifications
Specifications
Strep-tag II Peptide
-20°C
Strep-tag II Peptide
1.8 mg
Used for Competitive Elution of Strep-Tag II Proteins Bound to Strep MAB Classic Columns
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only.