Learn More
Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
Brand: Sampling Systems™ 8045R-125S
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged in ultra-strong packing with easy to open design
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Large variety of different sizes available
- Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
- Positioned the ergonomic handle high up on the scoop
- No Cross Contamination
- No Cleaning required
Specifications
Disposable Scoop | |
Individually Bagged | |
Single-use | |
Red |
Polystyrene | |
125 mL | |
Sterile |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
120313 | Certificate of Compliance | 11/03/2025 | 8045R-125S |
119268 | Certificate of Compliance | 19/08/2024 | 8045R-125S |
117292 | Certificate of Compliance | 09/02/2024 | 8045R-125S |
109371 | Certificate of Compliance | 09/08/2023 | 8045R-125S |
107697 | Certificate of Compliance | 30/04/2023 | 8045R-125S |