Learn More
Sampling Systems™ SteriWare™ Blue Disposable PharmaScoop™
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Blue Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
Brand: Sampling Systems™ 8045B-60S
This item is not returnable.
View return policy
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged in ultra-strong packing with easy to open design
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Large variety of different sizes available
- Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
- Positioned the ergonomic handle high up on the scoop
- No Cross Contamination
- No Cleaning required
Specifications
Disposable Scoop | |
Individually Bagged | |
Single-use | |
Blue |
Polypropylene | |
60 mL | |
Sterile |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
121315 | Certificate of Compliance | 19/03/2025 | 8045B-60S |
120341 | Certificate of Compliance | 11/02/2025 | 8045B-60S |
067348 | Certificate of Compliance | 13/01/2025 | 8045B-60S |
118368 | Certificate of Compliance | 02/12/2024 | 8045B-60S |
120339 | Certificate of Compliance | 20/11/2024 | 8045B-60S |
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Blue Disposable PharmaScoop™ > Capacity, Metric: 60mL