Learn More
Thermo Scientific™ Sterilin™ Cones
Safely mount your filter funnels without risk of chipping or slipping. Made of natural rubber, cones are also useful for mounting crucibles onto adapters.
Brand: Thermo Scientific™ CON0006
46.30 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Safely mount your filter funnels without risk of chipping or slipping. Thermo Scientific™ Sterilin™ Cones are made of natural rubber, and are also useful for mounting crucibles onto adapters.
- Set of six cones
- Manufactured from natural rubber
- For mounting filter funnels into Büchner flasks
- Also suitable for mounting Gooch crucibles onto adapters
- Not available in all geographic regions
Specifications
Set of 6 cones; Assorted | |
Rubber | |
Büchner flasks and Gooch crucibles | |
Set of six cones |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M548670 | Certificate of Compliance | 20/12/2024 | CON0006 |
M499210 | Certificate of Compliance | 03/12/2024 | CON0006 |
M451470 | Certificate of Compliance | 03/12/2024 | CON0006 |
M369230 | Certificate of Compliance | 03/12/2024 | CON0006 |
M490270 | Certificate of Compliance | 03/12/2024 | CON0006 |