Learn More
Thermo Scientific™ Sterilin™ BenchGuard
GREENER_CHOICE
Protect benches and other surfaces against liquid spills with the highly absorbent Thermo Scientific™ Sterilin™ BenchGuard.
Brand:  Thermo Scientific™ BG60E
 View more versions of this product
Product Code. 11709232
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Protect benches and other surfaces against liquid spills with the highly absorbent Thermo Scientific™ Sterilin™ BenchGuard. Absorbent liner also provides a cushion for glassware when accidentally dropped on benchtop.
  • Available in both 400mL/m2 and 800mL/m2absorption capacities
  • Plastic coated on one side to prevent soak through
  • Available in roll or sheet form
  • Rolls supplied in easy to use dispenser packs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bench Guard
1.607612 ft.
1.6076 ft.
BenchGuard Extra
800 mL/m2
0.6 m
0.49 m
50 x Sheets
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Sterilin™ BenchGuard >