Learn More
Linker Industrietechnik™ Stainless Steel Test Sieve, 200 mm OD, Nylon Sieve Skin
Constructed of durable stainless steel with a nylon sieve skin.
Brand:  Linker Industrietechnik™ 552.050.881-0112
 View more versions of this product
Product Code. 11745598
  • 211.00€ / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Description
  • Stainless steel ring construction
  • Nylon sieve cloth/skin
  • Available in a variety of sieve mesh sizes
Specifications
Specifications
112 μm
Stainless Steel
Nylon
200 mm
112 μm