Learn More
Amsta Rostfritt Stainless Steel Scoops
Buckets manufactured to handle chemicals, etc. Made of 0.8 mm acid-resistant stainless steel plate (EN1.4404).
Brand:  Amsta Rostfritt 517552
 View more versions of this product
Product Code. 11731776
  • 403.00€ / Each
Estimated Shipment: 08-08-2025
to see stock.
Explore more special offers
Description
Fully welded with soldered handles without slips for dirt hides.
Specifications
Specifications
Scoop
Stainless Steel
2 L
Product Title
Amsta Rostfritt Stainless Steel Scoops > Capacity: 2L