Learn More
BD Microlance™ Stainless Steel Needles
Color coded for easy recognition of the needle’s guage. BD™ Microlance™ Stainless Steel Needles feature thin walls for optimal flow and fit Luer slip syringes. These sterile, individually wrapped needles ensure more comfortable insertion with less risk for punching.
Brand:  BD 300700
 View more versions of this product
Product Code. 13217519
Description
Description
Unique slip angle and polishing method results in less patient discomfort at the injection site and less risk for punching.
  • Hypodermic needles with thin walls for optimal flow
  • Fits syringes connected to a Luer adapter or Luer Lock connector
  • Color coded for quick recognition of gauge for easy selection of appropriate flow rate
  • Sterile and individually wrapped for hygienic, single-use application
  • Stainless steel needles with epoxy resin adhesives
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel with Epoxy Resin Adhesives
100 individually wrapped needles, thin cannula wall
0.6 mm
Yes
Needle
Bevel
Sterile
Blue
23 G
For single use only
1.25 in.
30 mm
SZR -150, connected to a Luer adapter or Luer Lock connector
Documents
One moment while we fetch your results.
Product Title
BD Microlance™ Stainless Steel Needles > Needle Length/O.D.: 30mm/0.6mm