Learn More
asecos™ Stainless Steel Disposal Bin
The Stainless Steel Disposal Bin is ideal for the safe collection of solvent-soaked cleaning rags and other waste.
543.00€ - 707.00€
Specifications
Color Gray
Autoclavable Not Autoclavable
Material Stainless Steel
Product Type Replacement Trash Bin
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
17826436
View Documents
asecos™
8640
20 L
543.00€
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
17886576
View Documents
asecos™
8641
50 L
707.00€
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
Description
Description
  • Operation with foot pedal
  • With lid
Specifications
Specifications
Gray
Not Autoclavable
Stainless Steel
Replacement Trash Bin