Learn More
asecos™ Stainless Steel Disposal Bin
The Stainless Steel Disposal Bin is ideal for the safe collection of solvent-soaked cleaning rags and other waste.
543.00€ - 707.00€
Specifications
Color | Gray |
---|---|
Autoclavable | Not Autoclavable |
Material | Stainless Steel |
Product Type | Replacement Trash Bin |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17826436
|
asecos™
8640 |
20 L |
543.00€
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
17886576
|
asecos™
8641 |
50 L |
707.00€
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
Description
- Operation with foot pedal
- With lid
Specifications
Gray | |
Not Autoclavable | |
Stainless Steel | |
Replacement Trash Bin |