Learn More
JP Selecta™ Wire Baskets
Manufactured from AISI 304 stainless steel.
235.00€ - 307.00€
Specifications
Autoclavable | Autoclavable |
---|---|
Product Type | Basket |
Features | 4 Divider compartments basket |
Material | Stainless Steel |
Product Code | Brand | Diameter (Metric) Outside | Height (Metric) Exterior | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Height (Metric) Exterior | Price | Quantity & Availability | |||||
13403669
|
JP Selecta™
1000495 |
270 mm | 200 mm |
235.00€
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
12092754
|
JP Selecta™
1000496 |
360 mm | 280 mm |
307.00€
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
12022804
|
JP Selecta™
1000780 |
440 mm | 220 mm |
281.00€
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
Description
- With electro-bright finish
- Designed to fit Selecta autoclaves
- Available in a range of sizes
Specifications
Autoclavable | |
Basket | |
4 Divider compartments basket | |
Stainless Steel |
Product Title
JP Selecta™ Wire Baskets