missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach Lange™ Stablcal™ Stabilized Formazine Turbidity Standard 10.0 NTU
Hach Lange Stablcal Stabilized Formazine Turbidity Standard 10.0 NTU, Volume: 500mL
242.00€
Specifications
Type | Stablcal Stabilized Formazine Turbidity Standard |
---|---|
Turbidity | 10.0 NTU |
Description
Hach Lange Stablcal Stabilized Formazine Turbidity Standard 10.0 NTU, Volume: 500mL
Specifications
Stablcal Stabilized Formazine Turbidity Standard | |
10.0 NTU |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- StablCal solution 10 NTU 100ml bottle
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : methenamine, sodiumsulphate