missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach Lange™ Stablcal™ Stabilized Formazine Turbidity Standard 10.0 NTU
Hach Lange Stablcal Stabilized Formazine Turbidity Standard 10.0 NTU, Volume: 500mL
Brand: Hach Lange™ 2659942
This item is not returnable.
View return policy
Description
Hach Lange Stablcal Stabilized Formazine Turbidity Standard 10.0 NTU, Volume: 500mL
Specifications
Stablcal Stabilized Formazine Turbidity Standard | |
10.0 NTU | |
100 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- StablCal solution 10 NTU 100ml bottle
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : methenamine, sodiumsulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction