missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer SST™ II Advance Tubes

Product Code. p-7160278
Click to view available options
Closure Color:
Red
Yellow
Unit Size:
Pack of 100
Pack of 1000
This item is not returnable. View return policy

Product Code. 13107693

Brand: BD Vacutainer 367957

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contain an inert gel barrier that separates the serum from the blood clot during centrifugation, preventing contamination of the serum.

  • Contain an inert gel barrier that separates the serum from the blood clot during centrifugation, preventing contamination of the serum.
  • Stable barrier between serum and clotted blood during transportation and storage, leading to better analyte stability.
  • Routine analytes in clinical chemistry such as potassium are still stable after a week of storage at 2-8°C.
  • Clinical evaluation of special chemistries such as therapeutic drugs, proteins, peptides, steroids and vitamins demonstrates a high degree of stability.
  • Enable faster centrifugation times of 5 minutes at 3000 g.
  • Better sample quality.
  • Optimization of the work flow: short centrifugation time, sample processing and archiving in the primary tube.
  • No requirement for secondary tubes, reducing the possibility of misidentification.
  • Minimum recommended clotting time is 30 minutes.
  • Serum separation tubes should be gently inverted 180° and back 5-6 times.
TRUSTED_SUSTAINABILITY

Specifications

Additive Silica (Clot Activator), Gel Separator
Material PET
Closure Type Hemogard
Capacity (Metric) 3.5 mL
Closure Color Yellow
Tube Size 13 x 75 mm
For Use With (Application) Venous blood sampling, serum analysis
Label Type Paper
compliance-icons
Product Identifier
  • TUBE SST II PLH 13X75 3.5 PLBL CE GLD
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 2
Hazard Statement
  • H350-May cause cancer.
  • H361d-Suspected of damaging the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : toluene, quartz
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.