Learn More
Cytiva Spin Adapters for Gravity Columns
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
Brand: Cytiva 28-9232-45
This item is not returnable.
View return policy
Description
- Convenient adapters, enable spinning of all gravity columns.
Specifications
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns. | |
10 Pack | |
Spin Column Adapter |
Polypropylene | |
Convenient and rapid cleanup with high reproducibility for high throughput by running multiple samples in PD-10 desalting column for centrifugation | |
For PD-10 Desalting Column |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Cytiva Spin Adapters for Gravity Columns > Adapts PD-10 desalting columns