missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Transfertube™ Disposable Sampling Pipet and Dropper

An innovative device for transferring liquid and coring semi-solid samples with one hand

Brand:  Spectrum™ 190195P

260.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11760604

  • 386.00€ / Pack of 250
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The large aperture is particularly useful for working with particulates, suspensions, pastes, gels & creams. Disposable transferring reduces the risk of cross contamination associated with re-used pipettes and cutting instruments.

  • Capacity: 4mL
  • Length: 6.8 in. (175mm)
  • Shaft and plunger L x Dia.: 6 x 0.24 in. (153 x 6mm)
  • Finger ring I.D. x O.D.: 0.65 x 0.8 in. (16.5 x 20mm)
Applications:
  • General: Powders, viscous solutions, slurries, liquids
  • Microbiology: bacterial or viral colony on agar
  • Ecology and Industrial: Soil, mud, sludge, core samples, oils
  • Cosmetics: creams, gels, waxes, sampling
  • Food processing: pastes, creams, syrups, concentrates

Specifications

Specifications

Polypropylene
6 in.
15.24 cm
Single-use
For bacterial or viral colony on agar in microbiology, powders, viscous solutions, slurries, liquids, soil, mud, sludge, core samples
4 mL
Transfer Pipet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Spectrum™ Transfertube™ Disposable Sampling Pipet and Dropper > Dropping Pipet, Capacity: 4mL; 250/pack

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.