Learn More
Buerkle™ Spare Spray Heads for Buerkle™ Spray Bottle
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Description
- Manufactured from polypropylene and polyethylene
- Lever pump with adjustable nozzle
Specifications
Specifications
Capacity (Metric) | 250 mL |
Volume (Metric) Dispense | 1.2 mL/Stroke |
Closure Color | White |
Product Type | Lever Pump Sprayer with Adjustable Nozzle |
For Use With (Equipment) | Spray Bottle |
Closure Material | LDPE, Polypropylene, Stainless Steel |
Closure Type | Spray |
Through the Closure Dispensing | Yes |
Product Content Correction
Product Title
Buerkle™ Spare Spray Heads for Buerkle™ Spray Bottle >