Learn More
Witeg™ Soxhlet Extractor Bulb
79.90€ - 122.00€
Specifications
Product Type | Soxhlet Extractor Bulb |
---|---|
For Use With (Equipment) | Soxhlet Extraction Equipment |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11817700
|
Witeg™
2 212 125 |
125 mL |
122.00€
Each |
Estimated Shipment: 05-05-2025
Log in to see stock. |
||||||
11807700
|
Witeg™
2 212 060 |
60 mL |
79.90€
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
Specifications
Soxhlet Extractor Bulb | |
Soxhlet Extraction Equipment |
Product Title
Witeg™ Soxhlet Extractor Bulb