missing translation for 'onlineSavingsMsg'
Learn More
Sotalol hydrochloride, 98%, Thermo Scientific™ Product Code.: 15405949

Sotalol hydrochloride, 98%, Thermo Scientific™

Product Code. 15405949
100mg
Click to view available options
Quantity:
100mg
1g
5g
This item is not returnable. View return policy

Product Code. 15405949

Brand: Thermo Scientific Alfa Aesar J63772.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent beta adrenergic antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 µM.

Solubility
Soluble in phosphate buffered saline, DMSO, ethanol, water, and methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-24-0
Molecular Formula C12H21ClN2O3S
Molecular Weight (g/mol) 308.82
MDL Number MFCD00242937
InChI Key VIDRYROWYFWGSY-UHFFFAOYNA-N
Synonym sotalol hydrochloride, sotalol hcl, betapace, betapace af, sotacor, sotalex, berlex, mead johnson 1999, sorine
PubChem CID 66245
ChEBI CHEBI:9207
IUPAC Name hydrogen N-(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride
SMILES [H+].[Cl-].CC(C)NCC(O)C1=CC=C(NS(C)(=O)=O)C=C1

Specifications

Quantity 100mg
Merck Index 14,8728
Solubility Information Soluble in phosphate buffered saline,DMSO,ethanol,water,and methanol.
Formula Weight 308.82
Percent Purity 98%
Chemical Name or Material Sotalol hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW03K021 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog NumberJ63772.06,J63772.MC,J63772.03
Lot NumberT30K010 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog NumberJ63772.06,J63772.MC,J63772.03
Lot NumberN15I038 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ63772.06,J63772.MC,J63772.03
Lot NumberN16D030 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ63772.06,J63772.MC,J63772.03
Lot NumberZ03E042 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ63772.06,J63772.MC,J63772.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sotalol hydrochloride, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.