missing translation for 'onlineSavingsMsg'
Learn More
Sodium tetraethylborate, 97%, pure, Thermo Scientific Chemicals Product Code.: 10476131

Sodium tetraethylborate, 97%, pure, Thermo Scientific Chemicals

Product Code. 10476131
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10476131

Brand: Thermo Scientific Acros 363880010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15523-24-7
Molecular Formula C8H20BNa
Molecular Weight (g/mol) 150.04
MDL Number MFCD00061547
InChI Key SZSBMTRYJRHYNI-UHFFFAOYSA-N
Synonym sodium tetraethylborate, sodium tetraethylboranuide, borate 1-,tetraethyl-, sodium 1:1, sodium tetraethylborate 1-, et4bna, sodiotetraethylboron v, borate 1-, tetraethyl-, sodium, sodium tetraethylborate 1g
PubChem CID 23681030
IUPAC Name sodium;tetraethylboranuide
SMILES [B-](CC)(CC)(CC)CC.[Na+]

Specifications

Melting Point 140.0°C to 142.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: reacts
Formula Weight 150.04
Percent Purity 97%
Grade Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Sodium tetraethylborate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium tetraethylborate
Signal Word
  • Danger
Hazard Category
  • Pyrophoric solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P335-Brush off loose particles from skin.
  • P370+P378-In case of fire: Use to extinguish.
  • P422-Store contents under .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium tetraethylborate, 97%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.