missing translation for 'onlineSavingsMsg'
Learn More
Sodium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™ Product Code.: 15681340

Sodium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15681340 Shop All Honeywell Fluka Products
6 x 1 kg
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
50 kg
6 x 1 kg
6 x 2.5 kg
100 kg
This item is not returnable. View return policy

Product Code. 15681340

Brand: Honeywell-Fluka 134646X1KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C (literature)
CAS Max % 100.0000%
Quantity 6 x 1 kg
Assay Percent Range 99.0 to 100.5%
Linear Formula Na2SO4
IUPAC Name disodium;sulfate
Formula Weight 142.04
Grade puriss.
Packaging Poly Bottle
Density 2.68 g/mL at 25 °C(lit.)
Chemical Name or Material Sodium sulfate, Puriss., meets analytical specification of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.