missing translation for 'onlineSavingsMsg'
Learn More
Sodium, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11301428

Sodium, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11301428
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11301428

Brand: Thermo Scientific Alfa Aesar 010336.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-23-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]

Specifications

Color Silver to Gray
Appearance Solid in various forms
Density 0.968g/cm3
Assay Percent Range 99.95% (Metals basis)
Quantity 5 g
Odor Odorless
Vapor Pressure 1.59 hPa (1mm Hg)
Chemical Name or Material Sodium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP18L028 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog Number010336.03,010336.06
Lot NumberP18L026 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog Number010336.03,010336.06
Lot NumberN14L013 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog Number010336.03,010336.06
Lot NumberU08K026 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number010336.03,010336.06
Lot NumberS05K064 Certificate TypeCertificate of Analysis Date25/06/2024 Catalog Number010336.03,010336.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.