missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SNF5 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand: Novus Biologicals™ NBP1-90013PEP
This item is not returnable.
View return policy
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human SMARCB1. The SNF5 Recombinant Protein Antigen is derived from E. coli. The SNF5 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.This is a blocking peptide for NBP1-90013. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

Specifications
6598 | |
Chromatography | |
0.5mg/mL | |
PBS and 1M Urea, pH 7.4. | |
SMARCB1 | |
29kDa | |
0.1 mL | |
E.Coli | |
EKENSPEKFALKLCSELGLGGEFVTTIAYSIRGQLSWHQKTYAFSENPLPTVEIAIRNTGDADQWCPLLETLTDAEMEKKIRDQDRNTRRMRRLANTA |
Human | |
>80% | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
Unlabeled | |
SNF5 | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP1-90013. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ SNF5 Recombinant Protein Antigen > 0.1mL
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction