Learn More
Molecular Probes™ SlowFade™ Diamond Antifade Mountant with DAPI
SlowFade™ Diamond Antifade Mountant with DAPI
Brand:  Molecular Probes™ S36964
285.00 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
 View more versions of this product
Product Code. 15451244
  • 384.00€ / Pack of 5
Explore more special offers
Description
Show More
Specifications
Specifications
5 x 2 mL
Contains 5 dropper bottles contaning 2 mL reagent each.
• Store at 2-8°C or frozen at -5 to -30°C.
• Protect from light.
• Stable for at least 6 months after receiving.
Mounting Solution, Antifade Solution
Antifade Mountant
SlowFade™
5 x 2 mL
Documents
Manuals
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2892449 Certificate TypeCertificate of Analysis Date21/05/2024 Catalog NumberS36964
Lot Number2738243 Certificate TypeCertificate of Analysis Date05/10/2023 Catalog NumberS36964
Lot Number241130 Certificate TypeCertificate of Analysis Date07/07/2023 Catalog NumberS36964
Lot Number240395 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberS36964
Lot Number2607631 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog NumberS36964
Product Title
Molecular Probes™ SlowFade™ Diamond Antifade Mountant with DAPI > 5 x 2mL