missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Assistent™ Sloping Stem Glass Funnels

For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Karl Hecht™ Assistent™ Sloping Stem Glass Funnels are manufactured from clear soda-lime glass.

Brand:  Karl Hecht™ 44000048

 View more versions of this product

Product Code. 10083761

  • 22.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Slope angle of 60°
  • Available in a range of diameters
Specifications

Specifications

150mm
Glass
60°
Laboratory Applications
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.