Learn More
Epredia™ SlideMate™ AS Slide Printer
Reduce specimen identification errors while enhancing lean workflow with the Epredia™ SlideMate™ AS On-Demand Slide Printer.
10950.00€ - 21295.00€
Products 4
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
15488881
View Documents
Epredia™
A83900001
Slide Printer AS Basic
10950.00€
Each
15408891
View Documents
Epredia™
A83900002
Slide Printer AS Basic System
21295.00€
Each
15418891
View Documents
Epredia™
A83900004
Slide Printer AS On-Demand
14625.00€
Each
15451031
View accessory products View Documents
Epredia™
A83900005
Slide Printer AS On-Demand Sys
19595.00€
Each
Description
Description
The SlideMate AS promotes single piece workflow though unique features designed specifically for on-demand printing.
  • Fully integrated color touch-screen and simple software are easy to use
  • Built-in barcode scanner reads cassette information and imports the data to the slide label
  • Connects easily to other computers or Laboratory Information Systems
  • Prints directly onto the slide, eliminating the need for hand writing or labels
Documents
Product Certifications
Product Content Correction
Product Title
Epredia™ SlideMate™ AS Slide Printer