Learn More
Poulten Graf™ Single Use Luer-Tip Two-Part Syringe
Use these syringes for veterinary medicine applications. Poulten Graf™ Single Use Luer-Tip Two-Part Syringe are PVC-free and non-toxic, sterile packed and depatched in blister packs. The single-use syringes are extremely tight, and have a transparent, smooth operating cylinder.
Brand:  Poulten Graf™ 7 41006
 View more versions of this product
Product Code. 10076041
  • 99.95€ / Pack of 100
Explore more special offers
Description
Description
  • Produced in accordance with DIN 13098
  • Available in a variety of sizes, with and without needle
Specifications
Specifications
Autoclavable
Polypropylene
Luer-tip 2-part Syringe
100/Pk.
Luer Tip
Sterile
CE
Screen printed graduation
Single-use
10 to 12 mL
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Poulten Graf™ Single Use Luer-Tip Two-Part Syringe > Volume: 10 to 12mL