missing translation for 'onlineSavingsMsg'
Learn More
Silicone Viscosity Standard, 5mPas, Bookfield™ Product Code.: 10543108

Silicone Viscosity Standard, 5mPas, Bookfield™

Product Code. 10543108
500 mL
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy

Product Code. 10543108

Brand: Brookfield

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
A convenient, reliable means of checking the calibration of most Brookfield viscometers. These fluids are calibrated at 25°C in 500mL bottles and are supplied with instructions and calibration certificate. These Newtonian oils are ideal for the calibration of Brookfield viscometers and each one is traceable to NIST standards. Silicone oils should not be used in the paint and some food industries. Similarly silicone fluid should not be used for the calibration of glass viscometers due to coating of the viscometer walls.
  • Accuracy: ±1%
  • Excellent temperature stability
  • Recommended for use with Brookfield and most other rotational viscometers
  • Economical

Specifications

Packaging Bottle
Quantity 500 mL
Viscosity 5mPas
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicone Viscosity Standard, 5mPas, Bookfield™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.