Learn More
BRAND™ Unisilkon™ Silicone Paste
Low viscosity. For greasing ground joints.
Brand: BRAND™ 61605
35.24 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- Unisilkon™ TK M 1011
- For use with greasing ground joints
- Love viscosity
- Operating temperature from -40 to 160°C
- 50g
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13