Learn More
Watson-Marlow Tubing Silicone Double Segment Manifold Tubing
Silicone Double Segment Manifold Tubing is pump tubing tor 401U/D1, 401U/DM2 and 401U/DM3 manual/auto control ultra compact pumps.
Brand:  Watson-Marlow Tubing 983.0229.000
 View more versions of this product
Product Code. 17817973
  • 342.00€ / Pack of 6
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Description
  • 401U/D1, 401U/DM2 and 401U/DM3 Manual/auto Control Ultra Compact Pumps
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Double Segment Tubing
2.29 mm
1.5 ft.
0.03 in.
0.09 in.
Silicone
0.46 m
0.88 mm