Learn More
Product Code | Brand | Packaging | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Quantity | Price | Quantity & Availability | |||||
10773124
|
Fisher Chemical
S/0693/53 |
Plastic Powder Jar | 500 g |
78.95€
500g |
Estimated Shipment: 24-04-2025
Log in to see stock. |
|||||
10403652
|
Fisher Chemical
S/0693/60 |
Plastic Powder Jar | 1 kg |
120.00€
1kg |
In Stock
Log in to see stock. |
|||||
11438812
|
Fisher Chemical
S/0693/65 |
Plastic Keg | 5 kg |
289.00€
5kg |
In Stock
Log in to see stock. |
|||||
Description
Silica Gel 60 Angstrom Pore Size, 35-70 Micron, for Column Chromatography
Product Title
Silica Gel 60 Angstrom Pore Size, 35-70 Micron, for Column Chromatography, Fisher Chemical™