Learn More
Sheep anti-Rabbit IgG whole molecule, Biotin, Polyclonal, Secondary Antibody, Abnova™
Brand:  Abnova PAB10842.2mg
Product Code. 16273130
  • 255.00€ / 2mg
Explore more special offers
Specifications
Sheep anti-Rabbit IgG whole molecule
Polyclonal
Sheep anti-rabbit IgG recognizes rabbit IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin.
Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) sodium azide, 10mg/mL BSA (ImmunoglobulLyophilized from and Protease free))
Rabbit IgG whole molecule
2 mg
Rabbit
Lyophilized
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Biotin
The optimal working dilution should be determined by the end user.
Sheep
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Secondary
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sheep anti-Rabbit IgG whole molecule, Biotin, Polyclonal, Secondary Antibody, Abnova™ > 2mg; Biotin