Learn More
Epredia™ Peel-A-Way™ Disposable Embedding Molds
Promote rapid paraffin hardening with Epredia Peel-A-Way Disposable Embedding Molds.
Brand: Epredia™ 2219
192.43 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
This item is not returnable.
View return policy
Description
- Constructed of rigid transparent plastic.
Specifications
Embedding Mold | |
22 x 22 x 20 mm | |
Square |
Plastic | |
Single-use |
Product Content Correction
Product Title
Epredia™ Peel-A-Way™ Disposable Embedding Molds > 0.87 x 0.87 x 0.79 in. (22 x 22 x 20mm)