Learn More
VELP Scientifica SER 158 Series Automatic Solvent Extractor
Fully automatic and cloud-enabled hot solvent extractor, for safety, fast, accurate, and reliable extractable matter determination.
POA
Product Code | Brand | Dimensions (D x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (D x W x H) | Price | Quantity & Availability | |||||
17587566
|
VELP Scientifica
S303A0380 |
546 x 546 x 570 mm |
POA
Each |
N/A | |||||
15783619
|
VELP Scientifica
S303A0390 |
358 x 546 x 570 mm |
POA
Each |
N/A | |||||
Description
FAST EXTRACTION
The automatic extraction process completes fully unattended analysis according to the Randall method that is five times faster than traditional Soxhlet. The high-speed heating plates in glass-ceramic ensure fast solvent boiling.
MINIMUM SOLVENT CONSUMPTION
The patent pending titanium condensers guarantee the unparalleled solvent recovery of more than 90% that is collected in cooled recovery tank.
HIGH PRODUCTIVITY AND VERSATILITY
The powerful ControlPad is able to control up to 4 SER 158 units (3 or 6 positions) independently allowing to increase the number of units according to throughput requirement.
MAXIMUM SAFETY
- SolventXpress™ ensures zero exposure to solvents
- SafeEnd™ prevents the overheating and burning of soluble matter
- Remote analysis interruption and immediate notifications trough VELP Ermes
EASE OF USE
The SER 158 Series makes hot solvent extraction easy and fast:
- Preset library of methods and possibility to add new ones and mark preferred methods
- Multilingual support Balance connection for automatic result calculation
- Barcorde reader to reduce risk of errors
SMART INSTRUMENT
The SER 158 is designed to work unattended 24/7 with Load and Go technologies, minimizing necessary operator intervention. Thanks to VELP Ermes connection is possible to monitor the analysis and be notified, anytime, anywhere.
Specifications
Accelerated Solvent Extractor | |
115/230 V |