Learn More
SI Analytics™ Septum
SI Analytics™ Septum is an important part of the titration process and equiptment.
Brand:  SI Analytics™ 285221224
 View more versions of this product
Product Code. 10493354
  • 41.05€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For titration
  • Accessory piece
  • For Karl Fischer™ titrator
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Septa
18GL
Karl Fischer Titration Vessel TZ 1751 and TZ 1754
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SI Analytics™ Septum > For Use With: Karl Fischer Titration Vessel TZ 1751 and TZ 1754