missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sensititre™ Gram Negative MIC Plate
Utilize the latest antimicrobials, including Doripenem and Tigecycline, for accurate, economical results with Thermo Scientific™ Sensititre™ Gram Negative MIC Plates.
313.00€
Specifications
Species | Multiple |
---|---|
0.5 McFarland Standard | H2O (Part No. T3339) |
Antimicrobial Agents | Various |
Broth Type | MHB (Part No. T3462) |
Antibiotic Code | Various |
Description
Utilize the latest antimicrobials, including Doripenem and Tigecycline, for accurate, economical results with Thermo Scientific™ Sensititre™ Gram Negative MIC Plates.
Utilize the latest antimicrobials, including Doripenem and Tigecycline, for accurate, economical results with Sensititre Gram Negative MIC Plates.
- Multi-isolate format decreases cost per test
- Automated, semiautomated or manual read options are available for ultimate flexibility
- Extended shelf life and room-temperature storage simplify inventory control
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Multiple | |
Various | |
Various | |
Gram-negative Bacteria | |
35°C O2 | |
1 | |
Box | |
Autoread or manual | |
50 μL |
H2O (Part No. T3339) | |
MHB (Part No. T3462) | |
MIC Plate | |
Gram negative organisms: Enterobacteriaceae, Non-Enterobacteriaceae, Proteus sp. | |
18 hr | |
10 | |
10/Pk. | |
For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL |