Learn More
Thermo Scientific™ Sensititre™ Gram Negative MIC Plate
Utilize the latest antimicrobials, including Doripenem and Tigecycline, for accurate, economical results with Thermo Scientific™ Sensititre™ Gram Negative MIC Plates.
Brand:  Thermo Scientific™ YGN2F
302.04 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15334869
  • 313.00€ / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Utilize the latest antimicrobials, including Doripenem and Tigecycline, for accurate, economical results with Thermo Scientific™ Sensititre™ Gram Negative MIC Plates.
Utilize the latest antimicrobials, including Doripenem and Tigecycline, for accurate, economical results with Sensititre Gram Negative MIC Plates.
  • Multi-isolate format decreases cost per test
  • Automated, semiautomated or manual read options are available for ultimate flexibility
  • Extended shelf life and room-temperature storage simplify inventory control
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
H2O (Part No. T3339)
Multiple
Sensititre Gram Negative MIC Plate
MIC Plate
Minimum Inhibitory Concentration (MIC), Breakpoint
35°C O2
1
Box
Autoread or manual
50 μL
Various
MHB (Part No. T3462)
Various
Gram-negative Bacteria
Gram negative organisms: Enterobacteriaceae, Non-Enterobacteriaceae, Proteus sp.
18 hr
10
10/Pk.
For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL