missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Semi-Dry Blotters

Ideal for fast transfer of proteins and nucleic acids without the need for gel cassettes and tanks.

Brand:  Fisherbrand™ V10-SDB

 View more versions of this product

Product Code. 12388556

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Designed to produce an extremely uniform system for the transfer of proteins and nucleic acids.
  • Very low buffer volume and current, 0.8mA per cm2 of gel, produces very little heat, preventing band distortion
  • Transfers can be achieved in less than one hour, depending on the molecular weight of the molecules
  • Either single gels or multiple gel stacks can be used with the units
  • Colour coded electrodes: One electrode is manufactured from stainless steel and the other from platinised titanium, making them virtually corrosion free
  • Fast transfer times
  • Available in two sizes
 V10-SDB: 10 x 10cm format - accommodates one 10 x 10cm mini-gel
 V20-SDB: 20 x 20cm format - accommodates up to four 10 x 10cm mini-gels or one 20 x 20cm maxi-gel

Specifications

Specifications

Semi Dry Blotter
7.5 x 17 x 17 cm
80 mA / 0.8 mAcm-2
Active Transfer Area: L x W: 10 x 10 cm
Recommended Power Supplies: EV243
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions