Learn More
Secondary Drinking Water Metals, 4 Components, Honeywell Fluka™
Brand:  Honeywell Fluka E2165-100ML
Product Code. 16686083
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Secondary Drinking Water Metals
1.05 g/cm3
Standards for ICP and ICP-MS
100 mL
<2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Secondary Drinking Water Metals, 4 Components, Honeywell Fluka™ > 100 mL