Learn More
BOLA Scrapers
Scrapers PTFE, solid
157.00€ - 357.00€
Specifications
Sterility Non-sterile
Autoclavable Autoclavable
View More Specs
Products 3
Product Code Brand Width (Metric) Price Quantity & Availability  
Product Code Brand Width (Metric) Price Quantity & Availability  
10309381
View Documents
BOLA
H916-02
50 mm
157.00€
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
10610372
View Documents
BOLA
H916-06
90 mm
330.00€
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
10013551
View Documents
BOLA
H916-08
120 mm
357.00€
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Description
Description
  • Scrapers made of PTFE with tapered end.
  • Ideal handling due to big handle and wide blade.
  • Universal chemical resistance.
Specifications
Specifications
Non-sterile
Autoclavable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BOLA Scrapers